missing translation for 'onlineSavingsMsg'
Learn More

Tbj51 Antibody, Novus Biologicals™

Product Code. 18339471
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18339471 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18339471 Supplier Bio-Techne Supplier No. NBP318235

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Tbj51 Polyclonal antibody specifically detects Tbj51 in Protozoa samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tbj51
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS pH 7.4, 50% glycerol
Gene Alias Tb04.1H19.310, TPR-repeat-containing chaperone protein DNAJ
Host Species Rabbit
Immunogen Synthetic peptide from the C-terminal of Trypanosoma brucei brucei Tbj51 (aa. 605-705)
Purification Method Peptide affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3656749
Target Species Protozoa
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.