missing translation for 'onlineSavingsMsg'
Learn More

TbHsp70 Antibody, Novus Biologicals™

Product Code. 18325043 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18325043 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18325043 Supplier Novus Biologicals Supplier No. NBP318236

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TbHsp70 Polyclonal antibody specifically detects TbHsp70 in Protozoa samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TbHsp70
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS pH 7.4, 50% glycerol
Gene Alias heat shock 70 kDa protein 4
Host Species Rabbit
Immunogen Synthetic peptide from the mid-protein of Trypanosoma brucei brucei TbHsp70 (aa. 200-300)
Purification Method Peptide affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Protozoa
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.