missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress TBHBA

Product Code. 16479457
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16479457 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16479457 Supplier MedChemExpress Supplier No. HY15929
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress The Boehringer Mannheim cholesterol esterase/cholesterol oxidase/peroxidase/3,4-dichlorophenol kinetic reagent was modified by the inclusion of TBHBA (2,4,6-Tribromo-3-hydroxybenzoic acid) which reacts with hydrogen peroxide and 4-aminophenazone to produce a quinone-imine dye with a greater molar absorptivity than that produced with phenol.

Specifications

CAS 14348-40-4
Solubility DMSO : ≥ 39 mg/mL (104.05 mM)
Synonym 2,4,6-Tribromo-3-hydroxybenzoic acid
Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Quantity 5 g
Molecular Weight (g/mol) 374.81 g/mol
Product Type Biochemical Assay Reagents
Purity or Quality Grade 99.78%
Shipping Condition Room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.