missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals TBE urea sample buffer (2X)
Click to view available options
Quantity:
10 mL
25 mL
Unit Size:
10mL
25mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Sample Loading Buffer |
| Color | Blue |
| Concentration | 2X |
| Physical Form | Liquid |
| Quantity | 25 mL |
Safety and Handling
Product Identifier
- TBE urea sample buffer (2X)
- Danger
- Reproductive toxicity Category 1B
- H360FD-May damage fertility. May damage the unborn child.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Boric acid, phenol
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction