missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals TBE running buffer (10X)

Product Code. 15465089
Click to view available options
Quantity:
1 L
2 L
4 L
Unit Size:
1 L
2 L
4 L
This item is not returnable. View return policy

Product Code. 15465089

Brand: Thermo Scientific Chemicals J62788.K3

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Commonly used in nucleic acid electrophoresis. TBE Buffer is used for polyacrylamide and agarose gel electrophoresis. It is ideally suited for DNA and RNA separation on longer run, higher voltage or amperage gels. TBE is used with non-denaturing or denaturing (7 M urea) gels.

TBE (Tris/Borate/EDTA) Buffer is commonly used in nucleic acid electrophoresis. TBE Buffer is used for polyacrylamide and agarose gel electrophoresis. It is ideally suited for DNA and RNA separation on longer run, higher voltage or amperage gels. TBE is used with non-denaturing or denaturing (7 M urea) gels. It is also routinely used for DNA automated sequencing gel.5 TBE can also be used for agarose gels but is not recommended for preparative gels for recovery of nucleic acids. Tris-Borate-EDTA buffer has been used for pulsed-field gel electrophoresis (PFGE).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material TBE running buffer (10X)
Physical Form Liquid
Quantity 2 L
Packaging Plastic bottle
Solubility Information It is soluble in water.
My Thermo Scientific brand TBE buffer has precipitated out of solution. What can I do?

Concentrations of 5X Tris Borate EDTA (TBE) buffer or greater may precipitate due to the nature of the product. This precipitation is a result of nucleation of salt crystals by dust particles or other insoluble materials. In some cases, we recommend heating the buffer to ~ 50°C to re-dissolve the TBE into solution. Alternately, to avoid precipitation of high concentration TBE buffers, after the solution is prepared, you can filter it through a 0.2 µm cellulose acetate or cellulose nitrate filter. These treatments have no deleterious effect on the buffering properties of TBE.

compliance-icons compliance-icons
Product Identifier
  • TBE running buffer (10X)
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Boric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.