missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ TBCK Antibody, Novus Biologicals™

Product Code. 30044534 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30044534 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30044534 Supplier Novus Biologicals™ Supplier No. NBP329440

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TBCK Polyclonal antibody specifically detects TBCK in Human samples. It is validated for Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBCK
Applications Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation Tris-citrate/phosphate pH 7 -8.
Gene Alias HSPC302, MGC16169, TBC domain-containing protein kinase-like protein, TBC1 domain containing kinase, TBCKL
Host Species Rabbit
Immunogen Between 800 and 850
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 93627
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.