missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBCE Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555343
This item is not returnable.
View return policy
Description
Immunogen sequence: DWELKTQQPL MLKNQLLTLK IKYPHQLDQK VLEKQLPGSM TIQKVKGLLS RLLKVPVSDL LLSYESPKKP GREIELENDL KSLQFYSVEN GDCLLVRW Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 85%.
Cofactor E is one of four proteins (cofactors A, D, E, and C) involved in the pathway leading to correctly folded beta-tubulin from folding intermediates. Cofactors A and D are believed to play a role in capturing and stabilizing beta-tubulin intermediates in a quasi-native confirmation. Cofactor E binds to the cofactor D/beta-tubulin complex; interaction with cofactor C then causes the release of beta-tubulin polypeptides that are committed to the native state.
Specifications
| TBCE | |
| Polyclonal | |
| Unconjugated | |
| TBCE | |
| 2610206D02Rik; beta-tubulin cofactor E; C530005D02Rik; HRD; hypoparathyroidism, growth and mental retardation, and dysmorphism; hypothetical protein LOC664760; KCS; KCS1; pac2; pmn; Tbce; Tbce protein; tubulin folding cofactor E; tubulin-folding cofactor E; Tubulin-specific chaperone E; tubulin-specific chaperone E-like protein; Unknown (protein for MGC:133938); zgc:123075 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6905 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15813 | |
| TBCE | |
| Recombinant protein corresponding to Human TBCE. Recombinant protein control fragment (Product #RP-94105). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction