missing translation for 'onlineSavingsMsg'
Learn More

TBCE Antibody, Novus Biologicals™

Product Code. 18393869 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18393869 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18393869 Supplier Novus Biologicals Supplier No. H00006905B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

TBCE Polyclonal antibody specifically detects TBCE in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBCE
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. AAH08654
Gene Alias HRDTubulin-folding cofactor E, KCS1KCS, Kenny-Caffey syndrome, pac2, tubulin folding cofactor E, tubulin-specific chaperone e
Host Species Mouse
Immunogen TBCE (AAH08654, 1 a.a. - 527 a.a.) full-length human protein. MSDTLTADVIGRRVEVNGEHATVRFAGVVPPVAGPWLGVEWDNPERGKHDGSHEGTVYFKCRHPTGGSFIRPNKVNFGTDFLTAIKNRYVLEDGPEEDRKEQIVTIGNKPVETIGFDSIMKQQSQLSKLQEVSLRNCAVSCAGEKGGVAEACPNIRKVDLSKNLLSSWDEVIHIADQLRHLEVLNVSENKLKFPSGSVLTGTLSVLKVLVLNQTGITWAEVLRCVAGCPGLEELYLESNNIFISERPTDVLQTVKLLDLSSNQLIDENQLYLIAHLPRLEQLILSDTGISSLHFPDAGIGCKTSMFPSLKYLVVNDNQISQWSFFNELEKLPSLRALSCLRNPLTKEDKEAETARLLIIASIGQLKTLNKCEILPEERRRAELDYRKAFGNEWKQAGGHKDPEKNRLSEEFLTAHPRYQFLCLKYGAPEDWELKTQQPLMLKNQLLTLKIKYPHQLDQKVLEKQLPGSMTIQKVKGLLSRLLKVPVSDLLLSYESPKKPGREIELENDLKSLQFYSVENGDCLLVRW
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6905
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.