missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBCE Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£173.00 - £255.00
Specifications
| Antigen | TBCE |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1-5uL/mg of lysate, Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18642379
|
Novus Biologicals
NBP3-06359-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18641429
|
Novus Biologicals
NBP3-06359-50ul |
50 μg |
£173.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TBCE Polyclonal antibody specifically detects TBCE in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
| TBCE | |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Stem Cell Markers | |
| PBS, pH7.0 | |
| 6905 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-5uL/mg of lysate, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| HRDTubulin-folding cofactor E, KCS1KCS, Kenny-Caffey syndrome, pac2, tubulin folding cofactor E, tubulin-specific chaperone e | |
| Produced in rabbits immunized with E. coli-derived Human TBCE fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title