missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBCD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | TBCD |
|---|---|
| Concentration | 0.19 mg/mL |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16823744
|
Proteintech
14867-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16813744
|
Proteintech
14867-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cofactor D is one of four proteins (cofactors A, D, E, and C) involved in the pathway leading to correctly folded beta-tubulin from folding intermediates. Cofactors A and D are believed to play a role in capturing and stabilizing beta-tubulin intermediates in a quasi-native confirmation. Cofactor E binds to the cofactor D/beta-tubulin complex; interaction with cofactor C then causes the release of beta-tubulin polypeptides that are committed to the native state.Specifications
| TBCD | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat, Human | |
| Q8BYA0, Q9BTW9 | |
| 100361417, 108903, 6904 | |
| TBCD Fusion Protein Ag6310 | |
| Primary | |
| -20°C | |
| TBCD |
| 0.19 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Beta tubulin cofactor D, KIAA0988, SSD 1, SSD1, TBCD, tfcD, tubulin folding cofactor D, Tubulin specific chaperone D | |
| Tbcd | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title