missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ TBCD 293T Cell Transient Overexpression Lysate

Product Code. 18393159 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.01mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18393159 0.1 mL 0.01mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18393159 Supplier Novus Biologicals™ Supplier No. H00006904T02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Overexpression Lysate

TRUSTED_SUSTAINABILITY

Specifications

Quantity 0.1 mL
Content And Storage Store at -80°C. Avoid freeze-thaw cycles.
Target TBCD
Synonym Beta-tubulin cofactor D, KIAA0988, SSD1, SSD-1, TFCD, tubulin folding cofactor D, Tubulin-folding cofactor D, tubulin-specific chaperone d
Gene Symbol TBCD
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.