missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBCC Antibody (OTI1B2), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74465AF647
This item is not returnable.
View return policy
Description
TBCC Monoclonal specifically detects TBCC in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot.
Specifications
| TBCC | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TBCC | |
| Full length recombinant protein of human TBCC (NP_003183) produced in HEK293T cells. | |
| 0.1 mL | |
| 6903.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI1B2 | |
| Western Blot | |
| CFCtubulin-specific chaperone c, tubulin folding cofactor C, Tubulin-folding cofactor C | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction