missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBCA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588944
This item is not returnable.
View return policy
Description
Immunogen sequence: MADPRVRQIK IKTGVVKRLV KEKVMYEKEA KQQEEKIEKM RAEDGENYDI KKQAEILQES RMMIPDCQRR LEAAYLDLQR ILENEKDLEE AEEYKEARLV LDSVKLEA; Positive Samples: OVCAR3, HeLa, 293T, U-87MG, Mouse testis; Cellular Location: Cytoplasm, cytoskeleton.
The product of this gene is one of four proteins (cofactors A, D, E, and C) involved in the pathway leading to correctly folded beta-tubulin from folding intermediates. Cofactors A and D are believed to play a role in capturing and stabilizing beta-tubulin intermediates in a quasi-native confirmation. Cofactor E binds to the cofactor D/beta-tubulin complex; interaction with cofactor C then causes the release of beta-tubulin polypeptides that are committed to the native state.
Specifications
| TBCA | |
| Polyclonal | |
| Unconjugated | |
| TBCA | |
| CFA; chaperonin cofactor A; co-chaperonin associated with a- & b-tubulin; cofactor A; Tbca; Tbca13; TCP1-chaperonin cofactor A; tubulin cofactor a; tubulin folding cofactor A; tubulin-folding cofactor A; Tubulin-specific chaperone A | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 21371, 366995, 6902 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.21 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O75347, P48428, Q6PEC1 | |
| TBCA | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-108 of human TBCA (NP_0045981). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction