missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBCA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578188
This item is not returnable.
View return policy
Description
Positive Control: 293T, A431, HeLa, HepG2 Predicted Reactivity: Mouse (90%), Rat (89%), Bovine (85%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The product of this gene is one of four proteins (cofactors A, D, E, and C) involved in the pathway leading to correctly folded beta-tubulin from folding intermediates. Cofactors A and D are believed to play a role in capturing and stabilizing beta-tubulin intermediates in a quasi-native confirmation. Cofactor E binds to the cofactor D/beta-tubulin complex; interaction with cofactor C then causes the release of beta-tubulin polypeptides that are committed to the native state.
Specifications
| TBCA | |
| Polyclonal | |
| Unconjugated | |
| TBCA | |
| CFA; chaperonin cofactor A; co-chaperonin associated with a- & b-tubulin; cofactor A; Tbca; Tbca13; TCP1-chaperonin cofactor A; tubulin cofactor a; tubulin folding cofactor A; tubulin-folding cofactor A; Tubulin-specific chaperone A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6902 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.1 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| O75347 | |
| TBCA | |
| Recombinant protein encompassing a sequence within the center region of human TBCA. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction