missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBCA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00219-100ul
This item is not returnable.
View return policy
Description
TBCA Polyclonal antibody specifically detects TBCA in Human samples. It is validated for ELISA
Specifications
| TBCA | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| CFA, chaperonin cofactor A, TCP1-chaperonin cofactor A, tubulin folding cofactor A, Tubulin-folding cofactor A, tubulin-specific chaperone a | |
| Produced in rabbits immunized with purified, recombinant Human TBCA (Accession#: O75347; Met1-Ala108) | |
| 100 μg | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 6902 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction