missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TBC1D9 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA552305

Product Code. 15700261

  • £476.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: VGKLFVAQPA KEGGSGGSGP SCHQGIPGVL FPKKGPGQPY VVESVEPLPA SLAPDSEEHS LGGQMEDIKL EDSSPRDNGA CSSMLISDDD TKDDSSMSSY SVLSAGSHEE DKLHCEDI Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 83%.

TBC1D9 may act as a GTPase-activating protein for Rab family protein(s).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TBC1D9
Polyclonal
Unconjugated
TBC1D9
GRAMD9; KIAA0882; MDR1; TBC1 domain family member 9; TBC1 domain family member 9A; TBC1 domain family, member 9 (with GRAM domain); TBC1D9; TBC1D9A
Rabbit
Antigen affinity chromatography
RUO
23158
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q6ZT07
TBC1D9
Recombinant protein corresponding to Human TBC1D9. Recombinant protein control fragment (Product #RP-93714).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.