missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBC1D9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551370
This item is not returnable.
View return policy
Description
Immunogen sequence: GDSLINFREF VSGLSAACHG DLTEKLKLLY KMHVLPEPSS DQDEPDSAFE ATQYFFEDIT PECTHVVGLD SRSKQGADDG FVTVSLKPDK GKRANSQENR NYLRLW Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 92%.
TBC1D9 may act as a GTPase-activating protein for Rab family protein(s).
Specifications
| TBC1D9 | |
| Polyclonal | |
| Unconjugated | |
| TBC1D9 | |
| GRAMD9; KIAA0882; MDR1; TBC1 domain family member 9; TBC1 domain family member 9A; TBC1 domain family, member 9 (with GRAM domain); TBC1D9; TBC1D9A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23158 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6ZT07 | |
| TBC1D9 | |
| Recombinant protein corresponding to Human TBC1D9. Recombinant protein control fragment (Product #RP-93715). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction