missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBC1D8B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£231.00 - £470.00
Specifications
| Antigen | TBC1D8B |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18443701
|
Novus Biologicals
NBP1-82190-25ul |
25 μL |
£231.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18208125
|
Novus Biologicals
NBP1-82190 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TBC1D8B Polyclonal specifically detects TBC1D8B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| TBC1D8B | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 54885 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ASQDGNQCSVIIPLREVLAIDKTNDSSKSVIISIKGKTAFRFHEVKDFEQLVAKLRLRCGAASTQYHDISTELAISSESTEPSDNFEVQSLTSQRECSKTVNTEALMTVFHPQNLETLNSKMLKEKMKEQSWKILFAECGRGVSMFRT | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| RP11-321G1.1, TBC1 domain family, member 8B (with GRAM domain) | |
| TBC1D8B | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel