missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBC1D5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17078-1-AP-150UL
This item is not returnable.
View return policy
Description
TBC1 domain family member 5 (TBC1D5) may act as a GTPase-activating protein for Rab family protein(s).Specifications
| TBC1D5 | |
| Polyclonal | |
| Unconjugated | |
| TBC1D5 | |
| KIAA0210, TBC1 domain family member 5, TBC1 domain family, member 5, TBC1D5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 501088, 72238, 9779 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q80XQ2, Q92609 | |
| Tbc1d5 | |
| TBC1D5 Fusion Protein Ag10991 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction