missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBC1D4 [p Thr642] Antibody (JE60-97), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32749
This item is not returnable.
View return policy
Description
TBC1D4 Monoclonal antibody specifically detects TBC1D4 in Human,Mouse,Rat samples. It is validated for Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| TBC1D4 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100 | |
| Akt substrate of 160 kDa, AS160Acrg embryonic lethality minimal region ortholog, DKFZp779C0666, KIAA0603TBC (Tre-2, BUB2, CDC16) domain-containing protein, TBC1 domain family member 4, TBC1 domain family, member 4 | |
| Synthetic phosphopeptide corresponding to residues surrounding Thr642 of human TBC1D4 protein. (Uniprot: O60343) | |
| 100 μL | |
| Apoptosis, B Cell Development and Differentiation Markers, Diabetes Research, Lipid and Metabolism, Phospho Specific, Signal Transduction | |
| 9882 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| JE60-97 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction