missing translation for 'onlineSavingsMsg'
Learn More

TBC1D4 Antibody, DyLight 405, Novus Biologicals™

Product Code. 18419497 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18419497 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18419497 Supplier Novus Biologicals Supplier No. NBP281935V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TBC1D4 Polyclonal antibody specifically detects TBC1D4 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBC1D4
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias Akt substrate of 160 kDa, AS160Acrg embryonic lethality minimal region ortholog, DKFZp779C0666, KIAA0603TBC (Tre-2, BUB2, CDC16) domain-containing protein, TBC1 domain family member 4, TBC1 domain family, member 4
Host Species Rabbit
Immunogen TBC1D4 antibody was raised against an 19 amino acid peptide near the carboxy terminus of human TBC1D4 . The immunogen is located within amino acids 1190 - 1240 of TBC1D4. Amino Acid Squence: ETLEKLERANSQLKRQNMD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, B Cell Development and Differentiation Markers, Diabetes Research, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9882
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.