missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBC1D3C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | TBC1D3C |
|---|---|
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18690756
|
Novus Biologicals
NBP2-46751 |
0.1 mL |
£513.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18654006
|
Novus Biologicals
NBP2-46751-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TBC1D3C Polyclonal antibody specifically detects TBC1D3C in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| TBC1D3C | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Rab GTPase-Activating Protein PRC17, TBC1 Domain Family Member 3C, TBC1 Domain Family Member 3C/3D, TBC1 Domain Family Member 3C-Like Protein ENSP00000341742, TBC1 Domain Family, Member 3C, TBC1D3D | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: CWQAEHPAERVRSAFAAPSTDSDQGT | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2), 40% Glycerol | |
| 414060 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title