missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBC1D3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-86844
This item is not returnable.
View return policy
Description
TBC1D3 Polyclonal specifically detects TBC1D3 in Human samples. It is validated for Western Blot.
Specifications
| TBC1D3 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 84218 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| FLJ56187, FLJ58835, prostate cancer gene 17 protein, protein TRE17-alpha, TBC1 domain family member 3, TBC1 domain family, member 3F, TBC1D3, TBC1D3E | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of Human TBC1D3 | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu