missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBC1D23 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106439
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total TBC1D23.
Putative Rab GTPase-activating protein which plays a role in vesicular trafficking (PubMed:28823707). Involved in endosome-to-Golgi trafficking. Acts as a bridging protein by binding simultaneously to golgins, including GOLGA1 and GOLGA4, located at the trans-Golgi, and to the WASH complex, located on endosome-derived vesicles (PubMed:29084197, PubMed:29426865). Together with WDR11 complex facilitates the golgin-mediated capture of vesicles generated using AP-1 (PubMed:29426865). Plays a role in brain development, including in cortical neuron positioning. May also be important for neurite outgrowth, possibly through its involvement in membrane trafficking and cargo delivery, 2 processes that are essential for axonal and dendritic growth. May act as a general inhibitor of innate immunity signaling, strongly inhibiting multiple TLR and dectin/CLEC7A-signaling pathways. Does not alter initial activation events, but instead affects maintenance of inflammatory gene expression several hours after bacterial lipopolysaccharide (LPS) challenge. [UniProt].
Specifications
| TBC1D23 | |
| Polyclonal | |
| Unconjugated | |
| TBC1D23 | |
| 4930451A13Rik; AU015720; AU043671; AU043778; D030022P07Rik; HCV non-structural protein 4A-transactivated protein 1; HCV nonstructural protein 4A-transactivated protein 1; hypothetical protein LOC393459; NS4ATP1; RGD1307925; TBC1 domain family member 23; TBC1 domain family, member 23; TBC1D23; W51689; zgc:63756 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 304019, 55773, 67581 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8K0F1, Q9NUY8 | |
| TBC1D23 | |
| A synthesized peptide derived from human TBC1D23(Accession Q9NUY8), corresponding to amino acid residues G542-H592. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction