missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBC1D23 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £364.00
Specifications
| Antigen | TBC1D23 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
TBC1D23 Polyclonal antibody specifically detects TBC1D23 in Human, Mouse samples. It is validated for Western BlotSpecifications
| TBC1D23 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| DKFZp667G062, HCV non-structural protein 4A-transactivated protein 1, HCV nonstructural protein 4A-transactivated protein 1, TBC1 domain family member 23, TBC1 domain family, member 23 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 250-600 of human TBC1D23 (NP_001186127.1). SDSKEEVIKFLENTPSSLNIEDIEDLFSLAQYYCSKTPASFRKDNHHLFGSTLLGIKDDDADLSQALCLAISVSEILQANQLQGEGVRFFVVDCRPAEQYNAGHLSTAFHLDSDLMLQNPSEFAQSVKSLLEAQKQSIESGSIAGGEHLCFMGSGREEEDMYMNMVLAHFLQKNKEYVSIASGGFMALQQHLADINVDGPENGYGHWIASTSGSRSSINSVDGESPNGSSDRGMKSLVNKMTVALKTKSVNVREKVISFIENTSTPVDRMSFNLPWPDRSCTERHVSSSDRVGKPYRGVKPVFSIGDEEEYDTDEIDSSSMSDDDRKEVVNIQTWINKPDVKHHFPCKEVK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 55773 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title