missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ TBC1D22B 293T Cell Transient Overexpression Lysate

Product Code. 18382109 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.01mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18382109 0.1 mL 0.01mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18382109 Supplier Novus Biologicals™ Supplier No. H00055633T01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Overexpression Lysate

TRUSTED_SUSTAINABILITY

Specifications

Quantity 0.1 mL
Content And Storage Store at -80°C. Avoid freeze-thaw cycles.
Research Category Signal Transduction
Target TBC1D22B
Synonym C6orf197, chromosome 6 open reading frame 197, dJ744I24.2, DKFZp762J0110, FLJ20337, MGC125626, MGC125627, RP4-744I24.2, TBC1 domain family member 22B, TBC1 domain family, member 22B
Gene Symbol TBC1D22B
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.