missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBC1D21 Monoclonal Antibody (OTI1C7)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525726
This item is not returnable.
View return policy
Description
TBC1D21 Monoclonal Antibody for Western Blot, IHC (P)
TBC1D21 may act as a GTPase-activating protein for Rab family protein(s). May be involved in acrosome formation and cytoskeletal reorganization during spermiogenesis, possibly by regulating RAB3A activity.
Specifications
| TBC1D21 | |
| Monoclonal | |
| 0.46 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8IYX1 | |
| TBC1D21 | |
| Full length human recombinant protein of TBC1D21 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1C7 | |
| Unconjugated | |
| TBC1D21 | |
| male germ cell-specific expressed, containing a RabGAP domain; MgcRabGAP; TBC1 domain family member 21; TBC1 domain family, member 21; TBC1D21 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 161514 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction