missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBC1D2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556577
This item is not returnable.
View return policy
Description
Immunogen sequence: LLMDKNHAKQ QVICKLSEKV TQDFTHPPDQ SPLRPDAANR DFLSQQGKIE HLKDDMEAYR TQNCFLNSEI HQVTKIWRKV Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 74%.
Acts as GTPase-activating protein for RAB7A. Signal effector acting as a linker between RAC1 and RAB7A, leading to RAB7A inactivation and subsequent inhibition of cadherin degradation and reduced cell-cell adhesion.
Specifications
| TBC1D2 | |
| Polyclonal | |
| Unconjugated | |
| TBC1D2 | |
| A630005A06Rik; armus; BC022630; BC1D2; Gm1037; likely ortholog of H. sapiens TBC1 domain family, member 2 (TBC1D2); PARIS1; PARIS-1; PP8997; Prostate antigen recognized and identified by SEREX 1; TBC1 domain family member 2; TBC1 domain family member 2A; TBC1 domain family, member 2; TBC1 domain family, member 2A; TBC1D2; TBC1D2A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55357 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BYX2 | |
| TBC1D2 | |
| Recombinant protein corresponding to Human TBC1D2. Recombinant protein control fragment (Product #RP-92894). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction