missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBC1D15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553329
This item is not returnable.
View return policy
Description
Immunogen sequence: LRVLEKDAEV IVDWRPLDDA LDSSSILYAR KDSSSVVEWT QAPKERGHRG SEHLNSYEAE WDMVNTVSFK RKPHTNGDAP SHRNGKSKWS FLFSLTDLKS IKQNKEGMGW SYLVFCLKDD VVLPALHF Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 94%.
Acts as a regulator of transcriptional activation. Plays a role in the inducible expression of cytokine genes in T-cells, especially in the induction of the IL-2.
Specifications
| TBC1D15 | |
| Polyclonal | |
| Unconjugated | |
| TBC1D15 | |
| 4432405K22Rik; AU022358; AU023730; AW557774; GAP for RAB7; GTPase-activating protein RAB7; Ly6dl; lymphocyte antigen 6, locus D ligand; Rab7-GAP; TBC1 domain family member 15; TBC1 domain family, member 15; Tbc1d15; Tre2/Bub2/Cdc16 domain family member 15; Tre-2-budding uninhibited by benzimidazole-cell division cycle 16 domain, domain family member 15 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64786 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TC07 | |
| TBC1D15 | |
| Recombinant protein corresponding to Human TBC1D15. Recombinant protein control fragment (Product #RP-90740). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction