missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBC1D15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531803
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, HeLa, HepG2. Predicted reactivity: Mouse (88%), Chimpanzee (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Acts as a regulator of transcriptional activation. Plays a role in the inducible expression of cytokine genes in T-cells, especially in the induction of the IL-2.
Specifications
| TBC1D15 | |
| Polyclonal | |
| Unconjugated | |
| TBC1D15 | |
| 4432405K22Rik; AU022358; AU023730; AW557774; GAP for RAB7; GTPase-activating protein RAB7; Ly6dl; lymphocyte antigen 6, locus D ligand; Rab7-GAP; TBC1 domain family member 15; TBC1 domain family, member 15; Tbc1d15; Tre2/Bub2/Cdc16 domain family member 15; Tre-2-budding uninhibited by benzimidazole-cell division cycle 16 domain, domain family member 15 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64786, 66687 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8TC07, Q9CXF4 | |
| TBC1D15 | |
| Recombinant protein encompassing a sequence within the center region of human TBC1D15. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction