missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBC1D10B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106456
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total TB10B.
Acts as GTPase-activating protein for RAB3A, RAB22A, RAB27A, AND RAB35. Does not act on RAB2A and RAB6A.
Specifications
| TBC1D10B | |
| Polyclonal | |
| Unconjugated | |
| TBC1D10B | |
| 1110003P22Rik; C87963; EPI64B; FP2461; Protein wz3-85; Rab27A-GAPbeta; rab27A-GAP-beta; TBC1 domain family member 10B; TBC1 domain family, member 10B; TBC1D10B | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 26000, 365372, 68449 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q4KMP7, Q8BHL3 | |
| TBC1D10B | |
| A synthesized peptide derived from human TBC1D10B(Accession Q4KMP7), corresponding to amino acid residues P142-V192. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction