missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBC1D10A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA545123
This item is not returnable.
View return policy
Description
Peptide sequence: KAILDAEPGP RPALQPSPSI RLPLDAPLPG SKAKPKPPKQ AQKEQRKQMK Sequence homology: Guinea Pig: 100%; Human: 100%.
Acts as GTPase-activating protein for RAB27A, but not for RAB2A, RAB3A, nor RAB4A. [UniProt].
Specifications
| TBC1D10A | |
| Polyclonal | |
| Unconjugated | |
| TBC1D10A | |
| AC004997.C22.2; AI447804; dJ130H16.1; dJ130H16.2; EBP50-PDX interactor of 64 kDa; EBP50-PDZ interactor of 64 kD; EPI64; EPI64 protein; mFLJ00288; Rab27A-GAP-alpha; TBC1 domain family member 10A; TBC1 domain family, member 10A; TBC1D10; Tbc1d10a | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 83874 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9BXI6 | |
| TBC1D10A | |
| Synthetic peptide directed towards the C-terminal of human TBC1D10A (aa 391-440). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction