missing translation for 'onlineSavingsMsg'
Learn More

TBB, MedChemExpress

Product Code. 16486048
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16486048 10 mg 10mg
16482187 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16486048 Supplier Medchem Express Supplier No. HY14394

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TBB is a cell-permeable and ATP-competitive CK2 inhibitor with an IC50 of 0.15 μM for rat liver CK2.

TBB is a cell-permeable and ATP-competitive CK2 inhibitor with an IC50 of 0.15 μM for rat liver CK2.

Specifications

Chemical Name or Material TBB
CAS 17374-26-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C6HBr4N3
Quantity 10 mg
Synonym NSC 231634 Casein Kinase II Inhibitor I
Solubility Information DMSO : ≥ 100 mg/mL (230.04 mM)
SMILES BrC1=C2C(NN=N2)=C(Br)C(Br)=C1Br
Molecular Weight (g/mol) 434.71
Formula Weight 434.71
Percent Purity 99.31%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.