missing translation for 'onlineSavingsMsg'
Learn More

TBA-354, MedChemExpress

Product Code. 16485198
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16485198 10 mg 10mg
16447927 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16485198 Supplier Medchem Express Supplier No. HY12485

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TBA-354 is a potent anti-tuberculosis compound; maintains activity against Mycobacterium tuberculosis H37Rv isogenic monoresistant strains.

TBA-354 is a potent anti-tuberculosis compound

Specifications

Chemical Name or Material TBA-354
CAS 1257426-19-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H15F3N4O5
Quantity 10 mg
Solubility Information DMSO : ≥ 46 mg/mL (105.42 mM)
SMILES FC(F)(F)OC1=CC=C(C2=CC=C(CO[C@H]3CN4C(OC3)=NC([N+]([O-])=O)=C4)C=N2)C=C1
Molecular Weight (g/mol) 436.34
Formula Weight 436.34
Percent Purity 98.29%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.