missing translation for 'onlineSavingsMsg'
Learn More

Tazobactam, 94%

Product Code. 15447859
Change view
Click to view available options
Quantity:
1 g
Packaging:
Plastic bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15447859 1 g Plastic bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15447859 Supplier Thermo Scientific Alfa Aesar Supplier No. J66483.03
 Restricted Item
Estimated Shipment: 20-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 89786-04-9 | C10H12N4O5S | 300.29 g/mol

Tazobactam is a drug is used in conjunction with beta-lactamase susceptible penicillins to treat infections caused by beta-lactamase producing organisms. It acts as a beta lactamase inhibitor. It is used in combination with piperacillin to broaden the spectrum of piperacillin antibacterial action. It is used as antibiotic in the treatment of nosocomial pneumonia caused by Pseudomonas aeruginosa.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 89786-04-9
Melting Point 115°C to 145°C
Boiling Point 77°C
Flash Point >110°C (230°F)
Packaging Plastic bottle
Molecular Formula C10H12N4O5S
MDL Number MFCD00917472 MFCD00867002
Quantity 1 g
Synonym tazobactam, tazobactam acid, tazobactamum, zosyn, unii-se10g96m8w, ytr-830h, ytr 830h, chembl404, ytr830h, 2s,3s,5r-3-methyl-7-oxo-3-1h-1,2,3-triazol-1-ylmethyl-4-thia-1-azabicyclo 3.2.0 heptane-2-carboxylic acid 4,4-dioxide
Solubility Information Soluble in water
InChI Key LPQZKKCYTLCDGQ-WEDXCCLWSA-N
SMILES C[C@]1(CN2C=CN=N2)[C@@H](N2[C@@H](CC2=O)S1(=O)=O)C(O)=O
IUPAC Name (2S,3S,5R)-3-methyl-4,4,7-trioxo-3-[(1H-1,2,3-triazol-1-yl)methyl]-4λ⁶-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Molecular Weight (g/mol) 300.29
PubChem CID 123630
ChEBI CHEBI:9421
Formula Weight 300.29
Percent Purity 94%
Physical Form Powder
Chemical Name or Material Tazobactam
Show More Show Less
compliance-icons
Product Identifier
  • Tazobactam
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: Tazobactam

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.