missing translation for 'onlineSavingsMsg'
Learn More

Tazarotene, TRC

Product Code. 17961795
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
Unit Size:
10mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17961795 10 mg 10mg
17962005 50 mg 50mg
17993565 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17961795 Supplier Toronto Research Chemicals Supplier No. TRCT01005010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118292-40-3
Molecular Formula C21 H21 N O2 S
Molecular Weight (g/mol) 351.46
Synonym Tazarotene, 3-Pyridinecarboxylic acid, 6-[2-(3,4-dihydro-4,4-dimethyl-2H-1-benzothiopyran-6-yl)ethynyl]-, ethyl ester, 3-Pyridinecarboxylic acid, 6-[(3,4-dihydro-4,4-dimethyl-2H-1-benzothiopyran-6-yl)ethynyl]-, ethyl ester (9CI), 2H-1-Benzothiopyran, 3-pyridinecarboxylic acid deriv., AGN 190168, Acnitaz, Tazarotene, Tazorac, Zorac
IUPAC Name ethyl 6-[2-(4,4-dimethyl-2,3-dihydrothiochromen-6-yl)ethynyl]pyridine-3-carboxylate
SMILES CCOC(=O)c1ccc(nc1)C#Cc2ccc3SCCC(C)(C)c3c2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C21H21NO2S/c1-4-24-20(23)16-7-9-17(22-14-16)8-5-15-6-10-19-18(13-15)21(2,3)11-12-25-19/h6-7,9-10,13-14H,4,11-12H2,1-3H3
Formula Weight 351.1293
Chemical Name or Material Tazarotene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.