missing translation for 'onlineSavingsMsg'
Learn More

Tazarotene Thiochromane Dimer, TRC

Product Code. 30009216
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009216 25 mg 25mg
30008141 50 mg 50mg
30009601 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009216 Supplier Toronto Research Chemicals Supplier No. TRCT01005525MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1010805-73-8
Molecular Formula C26 H26 S2
Molecular Weight (g/mol) 402.6146
Synonym 6,6'-(1,3-Butadiyne-1,4-diyl)bis[3,4-dihydro-4,4-dimethyl-2H-1-benzothiopyran
IUPAC Name 6-[4-(4,4-dimethyl-2,3-dihydrothiochromen-6-yl)buta-1,3-diynyl]-4,4-dimethyl-2,3-dihydrothiochromene
SMILES CC1(C)CCSc2ccc(cc12)C#CC#Cc3ccc4SCCC(C)(C)c4c3

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C26H26S2/c1-25(2)13-15-27-23-11-9-19(17-21(23)25)7-5-6-8-20-10-12-24-22(18-20)26(3,4)14-16-28-24/h9-12,17-18H,13-16H2,1-4H3
Formula Weight 402.1476
Chemical Name or Material Tazarotene Thiochromane Dimer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.