missing translation for 'onlineSavingsMsg'
Learn More

Tazarotene, MedChemExpress

Product Code. p-200096849
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16486746

Brand: Medchem Express HY15388

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tazarotene (AGN 190168) is a selective retinoic acid receptor (RAR) agonist for the treatment of plaque psoriasis and acne vulgaris.

Tazarotene (AGN 190168) is a selective retinoic acid receptor (RAR) agonist for the treatment of plaque psoriasis and acne vulgaris.

Specifications

Chemical Name or Material Tazarotene
Physical Form Solid
CAS 118292-40-3
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C21H21NO2S
For Use With (Application) COVID-19-immunoregulation
Synonym AGN 190168
Solubility Information DMSO : 50 mg/mL (142.26 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=CC=C(C#CC2=CC=C3C(C(C)(C)CCS3)=C2)N=C1)OCC
Molecular Weight (g/mol) 351.46
Formula Weight 351.46
Percent Purity 99.83%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.