missing translation for 'onlineSavingsMsg'
Få mere at vide

Tazarotene, MedChemExpress

Produktkode 16486746
Encompass_Preferred
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
100 mg
Förpackningsstorlek
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkode Quantity unitSize
16486746 100 mg 100mg
16486478 10 mg 10mg
16482577 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Denne vare kan ikke returneres. Se returpolitik
Produktkode 16486746 missing translation for 'mfr' Medchem Express missing translation for 'supplierNo' HY15388

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolitik

MedChemExpress Tazarotene (AGN 190168) is a selective retinoic acid receptor (RAR) agonist for the treatment of plaque psoriasis and acne vulgaris.

Tazarotene (AGN 190168) is a selective retinoic acid receptor (RAR) agonist for the treatment of plaque psoriasis and acne vulgaris.

Tekniske data

Chemical Name or Material Tazarotene
Physical Form Solid
CAS 118292-40-3
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C21H21NO2S
For Use With (Application) COVID-19-immunoregulation
Synonym AGN 190168
Solubility Information DMSO : 50 mg/mL (142.26 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=CC=C(C#CC2=CC=C3C(C(C)(C)CCS3)=C2)N=C1)OCC
Molecular Weight (g/mol) 351.46
Formula Weight 351.46
Percent Purity 99.83%
Grade Research
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.