missing translation for 'onlineSavingsMsg'
Learn More

Taxol Resistant Associated Protein 3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18325103
Change view
Click to view available options
Quantity:
25 μg
50 μg
Unit Size:
25µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18325103 25 μg 25µg
18325563 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18325103 Supplier Novus Biologicals Supplier No. NBP31191125UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Taxol Resistant Associated Protein 3 Polyclonal antibody specifically detects Taxol Resistant Associated Protein 3 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Taxol Resistant Associated Protein 3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.5 - 2 ug/ml
Formulation PBS
Gene Alias TRAG3
Host Species Rabbit
Immunogen Synthetic peptide derived from the human Taxol Resistant Associated Protein 3 protein. (Uniprot: Q9Y5P2)
Purification Method Antigen Affinity-purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.