missing translation for 'onlineSavingsMsg'
Learn More
Learn More
taxilin gamma Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22330
This item is not returnable.
View return policy
Description
taxilin gamma Polyclonal specifically detects taxilin gamma in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| taxilin gamma | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| ELRG, factor inhibiting activating transcription factor 4 (ATF4)-mediated transcription, FIAT, lipopolysaccharide-specific response protein 5, taxilin gamma | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 55787 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg | |
| NP_060830.2 | |
| TXLNG | |
| The immunogen this antibody was made to, maps to a region between residue 475 to 525 of human Taxilin Gamma using the numbering given in entry NP_060830.2 (GeneID 55787). | |
| 100 μL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction