missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
taxilin gamma Polyclonal antibody specifically detects taxilin gamma in Human, Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | taxilin gamma |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 594 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | ELRG, factor inhibiting activating transcription factor 4 (ATF4)-mediated transcription, FIAT, lipopolysaccharide-specific response protein 5, taxilin gamma |
| Host Species | Rabbit |
| Immunogen | The immunogen this antibody was made to, maps to a region between residue 478 to 528 of human Taxilin Gamma using the numbering given in entry NP_060830.2 (GeneID 55787). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?