missing translation for 'onlineSavingsMsg'
Learn More

taxilin gamma Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18423998 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18423998 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18423998 Supplier Novus Biologicals Supplier No. NBP222329AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

taxilin gamma Polyclonal antibody specifically detects taxilin gamma in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen taxilin gamma
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias ELRG, factor inhibiting activating transcription factor 4 (ATF4)-mediated transcription, FIAT, lipopolysaccharide-specific response protein 5, taxilin gamma
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 25 to 50 of human Taxilin Gamma using the numbering given in entry NP_060830.2 (GeneID 55787).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55787
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.