missing translation for 'onlineSavingsMsg'
Learn More

Taxifolin, MedChemExpress

Product Code. p-200098701
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16478996

Brand: Medchem Express HYN0136

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Taxifolin ((+)-Dihydroquercetin) exhibits important anti-tyrosinase activity. Taxifolin exhibits significant inhibitory activity against collagenase with an IC50 value of 193.3 μM. Taxifolin is an important natural compound with antifibrotic activity. Taxifolin is a free radical scavenger with antioxidant capacity.

Taxifolin ((+)-Dihydroquercetin) exhibits important anti-tyrosinase activity. Taxifolin exhibits significant inhibitory activity against collagenase with an IC50 value of 193.3 μM. Taxifolin is an important natural compound with antifibrotic activity. Taxifolin is a free radical scavenger with antioxidant capacity.

Specifications

Chemical Name or Material Taxifolin
CAS 480-18-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C15H12O7
Quantity 100 mg
Synonym (+)-Dihydroquercetin (+)-Taxifolin
Solubility Information DMSO : ≥ 100 mg/mL (328.68 mM)
SMILES O=C1[C@H](O)[C@@H](C2=CC=C(O)C(O)=C2)OC3=CC(O)=CC(O)=C13
Molecular Weight (g/mol) 304.25
Formula Weight 304.25
Percent Purity 95.38%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.