missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAX1BP3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93739-0.02ml
This item is not returnable.
View return policy
Description
TAX1BP3 Polyclonal antibody specifically detects TAX1BP3 in Human, Mouse samples. It is validated for Western Blot
Specifications
| TAX1BP3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| 1G5, CaM kinase-like vesicle-associated, caM kinase-like vesicle-associated protein, MGC8407, VACAMKL, vesicle-associated calmodulin-binding protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-124 of human TAX1BP3 (NP_055419.1). MSYIPGQPVTAVVQRVEIHKLRQGENLILGFSIGGGIDQDPSQNPFSEDKTDKGIYVTRVSEGGPAEIAGLQIGDKIMQVNGWDMTMVTHDQARKRLTKRSEEVVRLLVTRQSLQKAVQQSMLS | |
| 0.02 mL | |
| Signal Transduction | |
| 30851 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction