missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TAX1BP1 Recombinant Rabbit Monoclonal Antibody (23GB2100), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553223
This item is not returnable.
View return policy
Description
This gene encodes a HTLV-1 tax1 binding protein. The encoded protein interacts with TNFAIP3, and inhibits TNF-induced apoptosis by mediating the TNFAIP3 anti-apoptotic activity. Degradation of this protein by caspase-3-like family proteins is associated with apoptosis induced by TNF. This protein may also have a role in the inhibition of inflammatory signaling pathways. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| TAX1BP1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q3UKC1, Q66HA4, Q86VP1 | |
| TAX1BP1 | |
| A synthesized peptide derived from human TRAF6BP (720-789AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2100 | |
| Unconjugated | |
| TAX1BP1 | |
| 1200003J11Rik; 1700069J21Rik; Aa1076; AA930106; CALCOCO3; D6Ertd404e; D6Ertd772e; liver regeneration-related protein 2; liver regeneration-related protein LRRG004; PRO0105; T6BP; Tax1 (human T cell leukemia virus type I) binding protein 1; Tax1 (human T-cell leukemia virus type I) binding protein 1; Tax1 binding protein 1; tax1-binding protein 1; Tax1-binding protein 1 homolog; TAX1BP1; TRAF6-binding protein; TRAF6-binding protein homolog; TXBP151 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 246244, 52440, 8887 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction