missing translation for 'onlineSavingsMsg'
Learn More

TAX1BP1 Rabbit anti-Human, Mouse, Rat, Clone: 7Y9W3, Novus Biologicals™

Product Code. 18384912 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18384912 100 μg 100µL
18088513 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18384912 Supplier Novus Biologicals Supplier No. NBP315794100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

TAX1BP1 Monoclonal antibody specifically detects TAX1BP1 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAX1BP1
Applications Western Blot
Classification Monoclonal
Clone 7Y9W3
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias T6BP, Tax1 (human T-cell leukemia virus type I) binding protein 1, tax1-binding protein 1, TRAF6-binding protein, TXBP151CALCOCO3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 690-789 of human TAX1BP1 (Q86VP1). DSEDSKEDENVPTAPDPPSQHLRGHGTGFCFDSSFDVHKKCPLCELMFPPNYDQSKFEEHVESHWKVCPMCSEQFPPDYDQQVFERHVQTHFDQNVLNFD
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 8887
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.