missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAX1BP1 Antibody (S07-6F6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19794-50ul
This item is not returnable.
View return policy
Description
TAX1BP1 Monoclonal antibody specifically detects TAX1BP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| TAX1BP1 | |
| Monoclonal | |
| 0.3 mg/mL | |
| Western Blot 1:1000, Immunohistochemistry 1:20, Immunohistochemistry-Paraffin | |
| T6BP, Tax1 (human T-cell leukemia virus type I) binding protein 1, tax1-binding protein 1, TRAF6-binding protein, TXBP151CALCOCO3 | |
| A synthetic peptide of human TAX1BP1 (Uniprot # Q86VP1) | |
| 50 μg | |
| Neuroscience | |
| 8887 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| S07-6F6 | |
| Unconjugated | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction