missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Tavolixizumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558738
This item is not returnable.
View return policy
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Tavolixizumab, also known as Tyenne® (tocilizumab-aazg), is a biosimilar to Actemra® (tocilizumab), an Interleukin-6 (IL-6) receptor antagonist. Tavolixizumab targets the IL-6 receptor, thereby inhibiting the IL-6 signaling pathway, which plays a crucial role in the inflammatory response associated with autoimmune diseases such as rheumatoid arthritis and systemic juvenile idiopathic arthritis. The biosimilar is available in various formulations, including prefilled syringes, pen injectors, and vials for subcutaneous and intravenous administration, providing versatile treatment options. Tavolixizumab has demonstrated efficacy and safety comparable to the reference product, with serious infections being one of the noted adverse effects. Synonyms for Tavolixizumab include tocilizumab-aazg and Tyenne.
Specifications
| Tavolixizumab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Human | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction