missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tautomycin, MedChemExpress
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
Description
Tautomycin, an antifungal antibiotic isolated from the bacterium Streptomyces verticillatus, is a potent and specific inhibitor of protein phosphatases 1 and 2A and induces contraction of smooth muscle under Ca2+-free conditions, with Kiapp values of 0.16 nM and 0.4 nM for PP1 and PP2A, respectively.
Specifications
Specifications
| Chemical Name or Material | Tautomycin |
| Physical Form | Solid |
| CAS | 109946-35-2 |
| Color | White |
| Quantity | 25 μg |
| Purity Grade Notes | Research |
| Molecular Formula | C41H66O13 |
| For Use With (Application) | COVID-19-immunoregulation |
| SMILES | C[C@@H]([C@]([C@H](CC1)C)([H])O[C@@]1(CC[C@H]2C)O[C@H]2CC[C@H](C)C(C)=O)CC[C@H](O)[C@H](C)C(C[C@@H](O)[C@H](OC)[C@@H](C(C)C)OC(C[C@H](C(C(O3)=O)=C(C3=O)C)O)=O)=O |
| Molecular Weight (g/mol) | 766.96 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction